Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS35 Recombinant Rabbit Monoclonal Antibody (JB33-82)
GREENER_CHOICE

Catalog No. PIMA534647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534647 Supplier Invitrogen™ Supplier No. MA534647
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: PC-3M, U937, SH-SY-5Y, human colon cancer tissue, human kidney tissue, human placenta tissue, mouse colon tissue, 293T.

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS35
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB33-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene VPS35
Gene Accession No. Q96QK1
Gene Alias AI647796; DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; maternal-embryonic 3; Mem3; PARK17; TCCCTA00141; vacuolar protein sorting 35; vacuolar protein sorting 35 homolog; vacuolar protein sorting 35 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35; VPS35 retromer complex component
Gene Symbols VPS35
Host Species Rabbit
Immunogen Recombinant protein within Human VPS35 aa 80-220.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25479, 55737
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.