Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS36 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550862

Catalog No. PA550862


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total VPS36 protein.

VPS36, VPS22, and VPS25 (Slagsvold et al., 2005 ; Kamura et al., 2001 ).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VPS36
Polyclonal
Unconjugated
VPS36
1700010A24Rik; 2210415M20Rik; 2810408E15Rik; C13orf9; CGI-145; EAP45; ELL-associated protein of 45 kDa; ESCRT-II complex subunit VPS36; RGD1309754; vacuolar protein sorting 36 (yeast); vacuolar protein sorting 36 homolog; vacuolar protein sorting 36 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 36; VPS36
Rabbit
Antigen affinity chromatography
RUO
51028
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86VN1
VPS36
Synthetic peptide of human vacuolar protein sorting 36 homolog (S. cerevisiae).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.