Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS37A Mouse anti-Human, Clone: 1F9A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | VPS37A |
---|---|
Clone | 1F9A4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6759
![]() ![]() |
Proteintech Group Inc
663271IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene belongs to the VPS37 family, and encodes a component of the ESCRT-I (endosomal sorting complex required for transport I) protein complex, required for the sorting of ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies. Expression of this gene is downregulated in hepatocellular carcinoma, and mutations in this gene are associated with autosomal recessive spastic paraplegia-53. A related pseudogene has been identified on chromosome 5. Alternatively spliced transcript variants have been found for this gene.Specifications
VPS37A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ESCRT I complex subunit VPS37A, HCRP1, hVps37A, PQBP2, VPS37A | |
VPS37A | |
IgG1 | |
Protein G | |
Antibody |
1F9A4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8NEZ2 | |
137492 | |
VPS37A Fusion Protein Ag19108 | |
Primary | |
-20°C | |
VPS37A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title