Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS37A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555182 Supplier Invitrogen™ Supplier No. PA555182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QHELSESCSA SALQARLKVA AHEAEEESDN IAEDFLEGKM EIDDFLSSFM EKRTICHCRR AKEEKLQQAI AMHSQFHAPL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene belongs to the VPS37 family, and encodes a component of the ESCRT-I (endosomal sorting complex required for transport I) protein complex, required for the sorting of ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies. Expression of this gene is downregulated in hepatocellular carcinoma, and mutations in this gene are associated with autosomal recessive spastic paraplegia-53. A related pseudogene has been identified on chromosome 5. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS37A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vps37a
Gene Accession No. Q8NEZ2
Gene Alias 2210018P21Rik; 4930592A21Rik; AW261445; D8Ertd531e; ESCRT-I complex subunit VPS37A; HCRP1; Hepatocellular carcinoma-related protein 1; hVps37A; polyglutamine binding protein 2; PQBP2; SPG53; vacuolar protein sorting 37 homolog A; vacuolar protein sorting 37 homolog A (S. cerevisiae); vacuolar protein sorting 37A; vacuolar protein sorting 37A (yeast); vacuolar protein sorting-associated protein 37A; Vps37A; VPS37A, ESCRT-I subunit
Gene Symbols Vps37a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS37A. Recombinant protein control fragment (Product #RP-93840).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.