Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS37B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558300 Supplier Invitrogen™ Supplier No. PA558300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSEARFAGLS LVQLNELLED EGQLTEMVQK MEETQNVQLN KEMTLASNRS LAEGNLLYQP QLDTLKARLT QKYQELQVLF EAYQIKKTKL DRQSSSASL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. May be involved in cell growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS37B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS37B
Gene Accession No. Q9H9H4
Gene Alias 2300007F24Rik; AI415429; BC026744; ESCRT-I complex subunit VPS37B; hVps37B; RGD1304576; vacuolar protein sorting 37 homolog B; vacuolar protein sorting 37 homolog B (S. cerevisiae); vacuolar protein sorting 37B; vacuolar protein sorting 37B (yeast); vacuolar protein sorting-associated protein 37B; Vps37b; VPS37B, ESCRT-I subunit
Gene Symbols VPS37B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS37B. Recombinant protein control fragment (Product #RP-102818).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.