Learn More
Invitrogen™ VPS37B Polyclonal Antibody

Description
VPS37B Polyclonal Antibody for Western Blot, IHC (P), IP
Specifications
Specifications
| Antigen | VPS37B |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | VPS37B |
| Gene Accession No. | Q9H9H4 |
| Gene Alias | 2300007F24Rik; AI415429; BC026744; ESCRT-I complex subunit VPS37B; hVps37B; RGD1304576; vacuolar protein sorting 37 homolog B; vacuolar protein sorting 37 homolog B (S. cerevisiae); vacuolar protein sorting 37B; vacuolar protein sorting 37B (yeast); vacuolar protein sorting-associated protein 37B; Vps37b; VPS37B, ESCRT-I subunit |
| Gene Symbols | VPS37B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.