Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS37B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548373
Description
Predicted to react with mouse and rat based on sequence homology.
Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. May be involved in cell growth and differentiation.
Specifications
VPS37B | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H9H4 | |
VPS37B | |
KLH conjugated synthetic peptide between 99-127 amino acids from the Central region of human VPS37B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
VPS37B | |
Vacuolar protein sorting-associated protein 37B, hVps37B, ESCRT-I complex subunit VPS37B, VPS37B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79720 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction