Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS37C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114096 Supplier Invitrogen™ Supplier No. PA5114096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQLEREMALA TNRSLAERNL EFQGPLEISR SNLSDRYQEL RKLVERCQEQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

VPS37C is a subunit of ESCRT-I pathway required for sorting ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS37C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VPS37C
Gene Accession No. A5D8V6
Gene Alias ESCRT-I complex subunit VPS37C; FLJ20847; hVps37C; PML39; vacuolar protein sorting 37 homolog C; vacuolar protein sorting 37C; vacuolar protein sorting-associated protein 37C; VPS37C; VPS37C, ESCRT-I subunit
Gene Symbols VPS37C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human VPS37C (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55048
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.