Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113176
Description
VPS39 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.
Specifications
| VPS39 | |
| Polyclonal | |
| Unconjugated | |
| VPS39 | |
| A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; Pldn; TLP; TRAP1-like protein; vacuolar protein sorting 39 (yeast); vacuolar protein sorting 39 homolog; vacuolar protein sorting 39 homolog (S. cerevisiae); Vam6; Vam6/Vps39-like protein; Vam6P; Vps39; VPS39 HOPS complex subunit; VPS39, HOPS complex subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23339 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96JC1 | |
| VPS39 | |
| Recombinant Human Vam6/Vps39-like protein (263-490AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction