Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554138 Supplier Invitrogen™ Supplier No. PA554138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPILLIHRIK EGMEIPNLRD SLVKILQDYN LQILLREGCK KILVADSLSL LKKMHRTQMK GVLVDEENIC ESCLSPILPS DAAKPFSVVV FHCRHMFHKE CLPMPSMNSA AQFCNICSAK NRGPGSAILE M Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 32%.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS41
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS41
Gene Accession No. P49754
Gene Alias AI317346; fa01f11; HVPS41; hVps41p; HVSP41; mVam2; RGD1560511; S53; si:ch211-146l10.2; vacuolar assembly protein 41; vacuolar protein sorting 41 (yeast); vacuolar protein sorting 41 homolog; vacuolar protein sorting 41 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 41 homolog; Vam2; VAM2 homolog; VPS41; VPS41 HOPS complex subunit; VPS41 subunit of HOPS complex; VPS41, HOPS complex subunit; wu:fa01f11
Gene Symbols VPS41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS41. Recombinant protein control fragment (Product #RP-92601).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.