Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS41 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554176
Description
Immunogen sequence: VLKPLIYEMI LHEFLESDYE GFATLIREWP GDLYNNSVIV QAVRDHLKKD SQNKTLLKTL AELYTYDKNY GNALEIYLTL RHKDVFQLIH KHNLFSSIKD KIVLLMDFDS EKAVDML Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known.
Specifications
VPS41 | |
Polyclonal | |
Unconjugated | |
VPS41 | |
AI317346; fa01f11; HVPS41; hVps41p; HVSP41; mVam2; RGD1560511; S53; si:ch211-146l10.2; vacuolar assembly protein 41; vacuolar protein sorting 41 (yeast); vacuolar protein sorting 41 homolog; vacuolar protein sorting 41 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 41 homolog; Vam2; VAM2 homolog; VPS41; VPS41 HOPS complex subunit; VPS41 subunit of HOPS complex; VPS41, HOPS complex subunit; wu:fa01f11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27072 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49754 | |
VPS41 | |
Recombinant protein corresponding to Human VPS41. Recombinant protein control fragment (Product #RP-92600). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction