Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS45 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | VPS45 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
VPS45 Polyclonal specifically detects VPS45 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VPS45 | |
Polyclonal | |
Rabbit | |
Human | |
H1, H1VPS45, hlVps45, h-vps45, leucocyte vacuolar protein sorting 45, vacuolar protein sorting 45 homolog (S. cerevisiae), vacuolar protein sorting 45A, vacuolar protein sorting 45A (yeast homolog), vacuolar protein sorting 45A (yeast), vacuolar protein sorting-associated protein 45, VPS45A, VPS45AVPS54A, VPS45B, VSP45, VSP45A | |
VPS45 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11311 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KETTGIVSMVYTQSEILQKEVYLFERIDSQNREIMKHLKAICFLRPTKENVDYIIQELRRPKYTIYFIYFSNVISKSDVKSLAEADEQEVVA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title