Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518215
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518215 Supplier Invitrogen™ Supplier No. PA518215
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene VPS45
Gene Accession No. Q9NRW7
Gene Alias AI462172; AW554165; H1; H1VPS45; HGNC:14579; hlVps45; h-VPS45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; SCN5; vacuolar protein sorting 45 (yeast); vacuolar protein sorting 45 homolog; vacuolar protein sorting 45 homolog (S. cerevisiae); vacuolar protein sorting 45A; vacuolar protein sorting protein 45; vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45; Vps45; VPS45A; VPS45B; VPS54A; VSP45; Vsp45a
Gene Symbols VPS45
Host Species Goat
Immunogen Synthetic peptide sequence (ESSQVTSRSASRR) corresponding to the C-terminus amino acids of VPS45 (aa 558-570).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11311
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.