Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555619 Supplier Thermo Scientific Supplier No. PA555619
Add to Cart
Add to Cart

VPS45 Polyclonal Antibody

VPS45 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS45
Gene Accession No. Q9NRW7
Gene Alias AI462172; AW554165; H1; H1VPS45; HGNC:14579; hlVps45; h-VPS45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; SCN5; vacuolar protein sorting 45 (yeast); vacuolar protein sorting 45 homolog; vacuolar protein sorting 45 homolog (S. cerevisiae); vacuolar protein sorting 45A; vacuolar protein sorting protein 45; vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45; Vps45; VPS45A; VPS45B; VPS54A; VSP45; Vsp45a
Gene Symbols VPS45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS45. Recombinant protein control fragment (Product #RP-94237).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11311
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.