Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS45A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2401 Supplier Bioss Supplier No. BS12777R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators.

Specifications

Antigen VPS45A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VPS45
Gene Accession No. Q9NRW7
Gene Alias AI462172; AW554165; H1; H1VPS45; HGNC:14579; hlVps45; h-VPS45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; SCN5; vacuolar protein sorting 45 (yeast); vacuolar protein sorting 45 homolog; vacuolar protein sorting 45 homolog (S. cerevisiae); vacuolar protein sorting 45A; vacuolar protein sorting protein 45; Vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45; VPS45; VPS45A; VPS45B; VPS54A; VSP45; VSP45A
Gene Symbols VPS45
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VPS45A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11311
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.