Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110573 Supplier Invitrogen™ Supplier No. PA5110573
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTTSTLQKAI DLVTKATEED KAKNYEEALR LYQHAVEYFL HAIKYEAHSD KAKESIRAKC VQYLDRAEKL KDYLRSKEKH GKKPVKENQS EGKGSDSDSE GDNPEKKKLQ EQLMGAVVME.

The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. The mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be really an yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 16; the gene for the other resides on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS4A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene VPS4A
Gene Accession No. Q793F9, Q8VEJ9, Q9UN37
Gene Alias 4930589C15Rik; AI325971; AW553189; hVPS4; Protein SKD2; SKD1; SKD1A; SKD1-homolog; SKD2; vacuolar protein sorting 4 homolog A; vacuolar protein sorting 4 homolog A (S. cerevisiae); vacuolar protein sorting 4a; vacuolar protein sorting 4a (yeast); vacuolar protein sorting factor 4A; vacuolar protein sorting protein 4a; vacuolar protein sorting-associated protein 4A; vacuolar sorting protein 4; VPS4; VPS4-1; VPS4A; vps4-A
Gene Symbols VPS4A
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116733, 246772, 27183
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.