Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS4A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | VPS4A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
VPS4A Polyclonal specifically detects VPS4A in Human samples. It is validated for Western Blot.Specifications
VPS4A | |
Polyclonal | |
Rabbit | |
Protein Phosphatase | |
FLJ22197, hVPS4, Protein SKD2, SKD1, SKD1A, SKD2, vacuolar protein sorting 4 homolog A (S. cerevisiae), vacuolar protein sorting 4A (yeast homolog), vacuolar protein sorting factor 4A, vacuolar protein sorting-associated protein 4A, vacuolar sorting protein 4, VPS4-1SKD1-homolog, VPS4vacuolar protein sorting 4A (yeast) | |
VPS4A | |
IgG | |
49 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9UN37 | |
27183 | |
Synthetic peptides corresponding to VPS4A(vacuolar protein sorting 4 homolog A (S. cerevisiae)) The peptide sequence was selected from the N terminal of VPS4A. Peptide sequence LRSKEKHGKKPVKENQSEGKGSDSDSEGDNPEKKKLQEQLMGAVVMEKPN. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title