Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS4B Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119118AF594
Description
VPS4B Polyclonal antibody specifically detects VPS4B in Human samples. It is validated for Western BlotSpecifications
VPS4B | |
Polyclonal | |
Western Blot | |
Cell migration-inducing gene 1 protein, Protein SKD1, SKD1B, SKD1cell migration-inducing 1, Suppressor of K(+) transport growth defect 1, suppressor of K+ transport defect 1, vacuolar protein sorting 4 homolog B (S. cerevisiae), vacuolar protein sorting 4B, vacuolar protein sorting 4B (yeast), vacuolar protein sorting-associated protein 4B, VPS42, VPS4-2 | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human vacuolar protein sorting 4 homolog B using the numbering given in entry NP_004860.2 (GeneID 9525). | |
0.1 mL | |
Autophagy, Microautophagy | |
9525 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction