Learn More
Invitrogen™ VPS4B Polyclonal Antibody

Description
Immunogen sequence: AQKPVKEGQP SPADEKGNDS DGEGESDDPE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.
Specifications
Specifications
| Antigen | VPS4B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | VPS4B |
| Gene Accession No. | O75351 |
| Gene Alias | 8030489C12Rik; cell migration-inducing 1; cell migration-inducing gene 1 protein; MIG1; Protein SKD1; SKD1; SKD1B; Suppressor of K(+) transport growth defect 1; suppressor of K+ transport defect 1; vacuolar protein sorting 4 homolog B; vacuolar protein sorting 4 homolog B (S. cerevisiae); vacuolar protein sorting 4B; vacuolar protein sorting 4b (yeast); vacuolar protein sorting-associated protein 4B; VPS42; VPS4-2; Vps4b |
| Gene Symbols | VPS4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.