Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS51 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282053AF700
This item is not returnable.
View return policy
Description
VPS51 Polyclonal antibody specifically detects VPS51 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
VPS51 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 700 | |
ANG2, ANG3, C11orf2, C11orf3, chromosome 11 open reading frame 2, FFR, protein fat-free homolog, vacuolar protein sorting 51 homolog (S. cerevisiae) | |
VPS51 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human VPS51. The immunogen is located within amino acids 620 - 670 of VPS51. Amino Acid Squence: LYEEGVRKAQSSDSSKRT | |
0.1 mL | |
Signal Transduction | |
738 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction