Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566395
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566395 Supplier Invitrogen™ Supplier No. PA566395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPPAPDVLEFT DHGGSGFVGG LCQVAAAYQE LFAAQGPAGA EKLAAFARQL GSRYFALVER RLAQEQGGGD NSLLVRALDR FH Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

Acts as component of the GARP complex that is involved in retrograde transport from early and late endosomes to the trans-Golgi network (TGN). The GARP complex is required for the maintenance of protein retrieval from endosomes to the TGN, acid hydrolase sorting, lysosome function, endosomal cholesterol traffic and autophagy. VPS51 participates in retrograde transport of acid hydrolase receptors, likely by promoting tethering and SNARE-dependent fusion of endosome-derived carriers to the TGN. Acts as component of the EARP complex that is involved in endocytic recycling. The EARP complex associates with Rab4-positive endosomes and promotes recycling of internalized transferrin receptor (TFRC) to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS51
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS51
Gene Accession No. Q9UID3
Gene Alias 1110014N23Rik; 3110057M17Rik; AI837850; ANG2; ANG3; Another new gene 2 protein; C11orf2; C11orf3; Ffr; PP5382; Protein fat-free homolog; vacuolar protein sorting 51 homolog; vacuolar protein sorting 51 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 51 homolog; VPS51; VPS51 GARP complex subunit; VPS51, GARP complex subunit
Gene Symbols VPS51
Host Species Rabbit
Immunogen Recombinant Human VPS51. Recombinant protein control fragment (Product #RP-107068).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.