Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560071 Supplier Thermo Scientific Supplier No. PA560071
Add to Cart
Add to Cart

VPS52 Polyclonal Antibody

VPS52 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS52
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS52
Gene Accession No. Q8N1B4
Gene Alias Are1; D130068D18; D430041K17Rik; dJ1033B10.5; DKFZp547I194; fb94c11; LOW QUALITY PROTEIN: vacuolar protein sorting-associated protein 52 homolog; RP5-1033B10; SAC2; SAC2 suppressor of actin mutations 2-like protein; SACM2L; tclw5; vacuolar protein sorting 52 (yeast); vacuolar protein sorting 52 homolog; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; vacuolar protein sorting-associated protein 52-like protein; Vacuolar protein sorting-associated protein 52-like protein-like protein; Vps52; VPS52 GARP complex subunit; VPS52 subunit of GARP complex; VPS52, GARP complex subunit; wu:fb94c11
Gene Symbols VPS52
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS52. Recombinant protein control fragment (Product #RP-99254).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.