Learn More
Invitrogen™ VPS53 Polyclonal Antibody

Description
A suggested positive control is 293 cell lysate. PA5-20549 can be used with blocking peptide PEP-0669.
Specifications
Specifications
| Antigen | VPS53 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Vps53 |
| Gene Accession No. | Q5VIR6, Q8CCB4 |
| Gene Alias | 2010002A08Rik; 2310040I21Rik; 3100002B05Rik; Are1; Hccs1; hVps53L; PCH2E; pp13624; RGD1311391; SAC2 suppressor of actin mutations 2-like protein; Sacm2l; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting 53 (yeast); vacuolar protein sorting 53 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; Vacuolar protein sorting-associated protein 53 homolog; Vps52; Vps53; VPS53 GARP complex subunit; VPS53, GARP complex subunit |
| Gene Symbols | Vps53 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.