Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571766
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571766 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571766 Supplier Invitrogen™ Supplier No. PA571766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS54 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS54
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Vps54
Gene Accession No. Q5SPW0, Q9P1Q0
Gene Alias Hcc8; Hepatocellular carcinoma protein 8; hVps54L; mSLP8; PPP1R164; protein phosphatase 1, regulatory subunit 164; SLP-8p; Tumor antigen HOM-HCC-8; tumor antigen SLP-8p; Tumor antigen SLP-8p homolog; vacuolar protein sorting 54 (yeast); vacuolar protein sorting 54 homolog; vacuolar protein sorting 54 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 54; VPS54; VPS54 GARP complex subunit; VPS54, GARP complex subunit; Vps54l; vps54-like protein; Vsp54; wr
Gene Symbols Vps54
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 465-491 amino acids from the Central region of human VPS54.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245944, 51542
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.