Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566792 Supplier Invitrogen™ Supplier No. PA566792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKTAGNRLSGL LEAEEEDEFY QTTYGGFTEE SGDDEYQGDQ SDTEDEVDSD FDIDEGDEPS SDGEAEE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Vacuolar protein sorting-associated protein 72 homolog (VPS72), also known as VPS72 or Transcription factor-like 1, is a 364 amino acid subunit of the TRRAP/TIP60 HAT complex. VPS72 has also been identified as a subunit of a novel complex containing SNF2-related helicase SRCAP (SWI2/SNF2-related CBP activator protein). This SRCAP-containing complex is very similar to the S. cerevisiae SWR1 chromatin remodeling complex. The involvement of VPS72 in these complexes has suggested that VPS72 plays multiple roles in chromatin modification and remodeling in cells. VPS72 localizes to the nucleus and is phosphorylated upon DNA damage, most likely by ATM or ATR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS72
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VPS72
Gene Accession No. Q15906
Gene Alias CFL1; Protein YL-1; Swc2; T YL-1; TCFL1; Transcription factor-like 1; transformation suppressor gene YL-1; vacuolar protein sorting 72 (yeast); vacuolar protein sorting 72 homolog; vacuolar protein sorting 72 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 72 homolog; VPS72; Yl1; YL-1
Gene Symbols VPS72
Host Species Rabbit
Immunogen Recombinant Human VPS72. Recombinant protein control fragment (Product #RP-107449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6944
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.