Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VPS72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599249
Description
Antibody detects endogenous levels of total VPS72.
Vacuolar protein sorting-associated protein 72 homolog (VPS72), also known as VPS72 or Transcription factor-like 1, is a 364 amino acid subunit of the TRRAP/TIP60 HAT complex. VPS72 has also been identified as a subunit of a novel complex containing SNF2-related helicase SRCAP (SWI2/SNF2-related CBP activator protein). This SRCAP-containing complex is very similar to the S. cerevisiae SWR1 chromatin remodeling complex. The involvement of VPS72 in these complexes has suggested that VPS72 plays multiple roles in chromatin modification and remodeling in cells. VPS72 localizes to the nucleus and is phosphorylated upon DNA damage, most likely by ATM or ATR.
Specifications
VPS72 | |
Polyclonal | |
Unconjugated | |
VPS72 | |
CFL1; Protein YL-1; Swc2; T YL-1; TCFL1; Transcription factor-like 1; transformation suppressor gene YL-1; vacuolar protein sorting 72 (yeast); vacuolar protein sorting 72 homolog; vacuolar protein sorting 72 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 72 homolog; VPS72; Yl1; YL-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21427, 310661, 6944 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15906, Q62481 | |
VPS72 | |
A synthesized peptide derived from human VPS72(Accession Q15906), corresponding to amino acid residues S129-T179. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction