Learn More
Invitrogen™ VPS8 Monoclonal Antibody (2C8)

Description
Peptide Sequence: RITPSQVKMS PSYHQSKGDP TAKKGTSEPV LDPQQIQAFD QLCRLYRGSS RLALLTELSQ NRSSESYRPF SGSQSAPAFN SIFQNENFQL QLIPPPVTED.
Specifications
Specifications
| Antigen | VPS8 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2C8 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | VPS8 |
| Gene Accession No. | Q8N3P4 |
| Gene Alias | AI315068; AU040738; im:7166005; KIAA0804; mKIAA0804; RGD1309443; si:dkey-24b15.1; vacuolar protein sorting 8 homolog; vacuolar protein sorting 8 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 8 homolog; VPS8; VPS8 CORVET complex subunit; VPS8 subunit of CORVET complex; VPS8, CORVET complex subunit; zgc:158695 |
| Gene Symbols | VPS8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.