Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ vpx Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144354 Supplier Invitrogen™ Supplier No. PA5144354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

vpx Polyclonal Antibody for Western Blot, ELISA

Plays a role in nuclear translocation of the viral pre-integration complex (PIC), thus is required for the virus to infect non-dividing cells. Targets specific host proteins for degradation by the 26S proteasome. Acts by associating with the cellular CUL4A-DDB1 E3 ligase complex through direct interaction with host VPRPB/DCAF-1. This change in the E3 ligase substrate specificity results in the degradation of host SAMHD1. In turn, SAMHD1 depletion allows viral replication in host myeloid cells by preventing SAMHD1-mediated hydrolysis of intracellular dNTPs necessary for reverse transcription (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen vpx
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene HIV2gp3
Gene Accession No. P06939
Gene Alias Viral protein X; vpxProtein Vpx; X ORF protein
Gene Symbols HIV2gp3
Host Species Rabbit
Immunogen Recombinant Human immunodeficiency virus type 2 subtype A Protein Vpx protein (1-112AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1724714
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.