Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VR1 C-terminus (TRPV1) Guinea Pig anti-Human, Rat, Polyclonal, Neuromics™
Guinea Pig Polyclonal Antibody
Supplier: Neuromics, Inc GP1410050UL

Description
VR1 C-terminus (TRPV1) Polyclonal antibody specifically detects VR1 C-terminus (TRPV1) in Human, Rat samples. It is validated for ImmunohistochemistrySpecifications
VR1 C-terminus (TRPV1) | |
Polyclonal | |
Unconjugated | |
TRPV1 | |
Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; Vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; VR1; VR-1; Vr1l; Vr1l1 | |
YTGSLKPEDAEVFKDSMVPGEK | |
RUO | |
7442, 83810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Conc. Not Determined | |
whole serum with 0.02% sodium azide | |
O35433, Q8NER1 | |
Guinea Pig | |
50 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction