Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VR1 N-Terminus (TRPV1) Rabbit anti-Rat, Polyclonal, Neuromics™

Catalog No. 502100033
Encompass
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0033 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-210-0033 Supplier Neuromics, Inc Supplier No. RA1011050UL

Rabbit Polyclonal Antibody

VR1 N-Terminus (TRPV1) Polyclonal antibody specifically detects VR1 N-Terminus (TRPV1) in Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot

Specifications

Antigen VR1 N-Terminus (TRPV1)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene TRPV1
Gene Accession No. O35433
Gene Alias Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; Vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; VR1; VR-1; Vr1l; Vr1l1
Host Species Rabbit
Immunogen RASLDSEESESPPQENSC
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83810
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.