Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VRK1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299819IR
Description
VRK1 Polyclonal specifically detects VRK1 in Human samples. It is validated for Western Blot, ELISA.Specifications
VRK1 | |
Polyclonal | |
Western Blot, ELISA | |
EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1 | |
Produced in rabbits immunized with purified, recombinant VRK1 (Q99986; Met1-Lys396). | |
0.1 mL | |
Core ESC Like Genes, Protein Kinase, Stem Cell Markers | |
7443 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction