Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VRK1 Antibody (OTI2C9), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274855R
Description
VRK1 Monoclonal specifically detects VRK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VRK1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VRK1 | |
Recombinant protein fragment corresponding to amino acids 1-230 of human VRK1 (NP_003375) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI2C9 | |
Western Blot | |
EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Protein Kinase, Stem Cell Markers | |
7443.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction