Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VRK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581887 Supplier Invitrogen™ Supplier No. PA581887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAAQAGRQSS AKRHLAEQFA VGEIITDMAK KEWKVGLPIG QGGFGCIYLA DMNSSESVGS DAPCVVKVEP SDNGPLFTEL KFYQRAAKPE QIQKWIRTRK LKYLGVPKYW GSGLHDKNGK SYRFMIMDRF GSDLQKI Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 92%.

Human vaccinia-related kinase 1 (VRK1) is a new kinase related to a poxvirus kinase, and more distantly to the casein kinase 1 family. VRK1 shows strong autophosphorylating activity in several Ser and Thr residues. VRK has also been found to be an upstream regulator of p53 that belongs to a new signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VRK1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VRK1
Gene Accession No. Q99986
Gene Alias 51PK; MGC117401; MGC138280; PCH1; PCH1A; Serine/threonine-protein kinase 51PK; serine/threonine-protein kinase VRK1; vaccinia related kinase 1; vaccinia virus B1R-related kinase 1; vaccinia-related kinase 1; VRK1
Gene Symbols VRK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VRK1. Recombinant protein control fragment (Product #RP-88905).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.