Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VRK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587084
Description
Purity is > 95% by SDS-PAGE.
Human vaccinia-related kinase 1 (VRK1) is a new kinase related to a poxvirus kinase, and more distantly to the casein kinase 1 family. VRK1 shows strong autophosphorylating activity in several Ser and Thr residues. VRK has also been found to be an upstream regulator of p53 that belongs to a new signaling pathway.
Specifications
VRK1 | |
Polyclonal | |
Unconjugated | |
VRK1 | |
51PK; MGC117401; MGC138280; PCH1; PCH1A; Serine/threonine-protein kinase 51PK; serine/threonine-protein kinase VRK1; vaccinia related kinase 1; vaccinia virus B1R-related kinase 1; vaccinia-related kinase 1; VRK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22367, 362779, 7443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q80X41, Q99986 | |
VRK1 | |
Recombinant protein of human VRK1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction