Learn More
Promega VRK2 Kinase Enzyme System, 10microg
Catalog No. PRV4494 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4494 10 μg
1 options
Catalog No. PRV4494 Supplier Promega Supplier No. V4494
Recombinant human VRK2 (1-375) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. VRK2 (also known as vaccinia related kinase 2) is a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases.
  • Easily Screen and Profile VRK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C