Learn More
VRK2 Mouse anti-Human, Clone: OTI8H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811402

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, leukocytes, fetal liver, and carcinomas. Its protein localizes to the endoplasmic reticulum and has been shown to phosphorylate casein and undergo autophosphorylation. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
VRK2 | |
Monoclonal | |
Unconjugated | |
VRK2 | |
vaccinia related kinase 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86Y07 | |
VRK2 | |
Human recombit protein fragment corresponding to amino acids 301-486 of human VRK2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.