Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VRK2 Polyclonal Antibody

Catalog No. PIPA588440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588440 Supplier Invitrogen™ Supplier No. PA588440

Rabbit Polyclonal Antibody

Immunogen sequence: DELPQSVLKW APSGSSCCEI AQFLVCAHSL AYDEKPNYQA LKKILNPHGI PLGPLDFSTK GQSINVHTPN SQKVDSQKAA TKQVNKAHNR LIEKKVHSER SAESCATWKV QKEEKLIGLM NNEAAQESTR RRQKYQESQE PLNEVNSFPQ KISYTQFPNS FYEPHQDFTS PDIFKKSRSP SWYKYTSTVS TGITDLESST GLWPTISQFT LSEETNADVY YYRIIIPVLL MLVFLALFFL; Positive Samples: LO2; Cellular Location: Cytoplasm, Endoplasmic reticulum membrane, Mitochondrion membrane, Nucleus, Single-pass type IV membrane protein.

VRK2 serine/threonine kinases vaccinia related kinase 2 is a member of the vaccinia-related kinase (VRK) family of kinases. VRK2 is widely expressed in human tissues and highly expressed in actively dividing cells, such as those in testis, leukocytes, fetal liver and carcinomas. VRK2 can be used to phosphorylate casein and itself undergoes autophosphorylation. VRK2 interacts specifically with Epstein-Barr virus BHRF1, a homologue of Bcl-2, and this interaction is involved in protecting cells from apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VRK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VRK2
Gene Accession No. Q86Y07
Gene Alias 2810003O05Rik; AI447698; Serine/threonine-protein kinase VRK2; vaccinia related kinase 2; vaccinia virus B1R-related kinase 2; vaccinia virus BIR kinase related kinase 2; vaccinia-related kinase 2; Vrk2
Gene Symbols VRK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 269-508 of human VRK2 (NP_0062872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7444
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.