Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VRK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | VRK3 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
VRK3 Polyclonal specifically detects VRK3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VRK3 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
inactive serine/threonine-protein kinase VRK3, serine/threonine-protein kinase VRK3, Serine/threonine-protein pseudokinase VRK3, vaccinia related kinase 3, Vaccinia-related kinase 3 | |
VRK3 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
51231 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CPYCGNSLPVEEHVGSQTFVNPHVSSFQGSKRGLNSSFETSPKKVKWSSTVTSPRLSLFSDGDSSESEDTLSS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title