Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VRK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566016
Description
Immunogen sequence: CPYCGNSLPVE EHVGSQTFVN PHVSSFQGSK RGLNSSFETS PKKVKWSSTV TSPRLSLFSD GDSSESEDTL SS Highest antigen sequence identity to the following orthologs - mouse 68%, rat 73%.
This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms.
Specifications
VRK3 | |
Polyclonal | |
Unconjugated | |
VRK3 | |
Inactive serine/threonine-protein kinase VRK3; serine/threonine-protein kinase VRK3; serine/threonine-protein pseudokinase VRK3; vaccinia related kinase 3; vaccinia-related kinase 3; VRK3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV63 | |
VRK3 | |
Recombinant Human VRK3. Recombinant protein control fragment (Product #RP-106702). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction