Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VRL1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100139
Description
VRL1 Polyclonal specifically detects VRL1 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
VRL1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Osm-9-like TRP channel 2, transient receptor potential cation channel, subfamily V, member 2, TrpV2, Vanilloid receptor-like protein 1, VRL-1MGC12549, VRL1transient receptor potential cation channel subfamily V member 2, VRLOTRPC2 | |
Goat | |
86 kDa | |
0.1 mg | |
Primary | |
Rat. Predicted cross-reactivity based on sequence identity: Mouse. | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:4000 | |
Q9WTR1 | |
TRPV2 | |
Peptide with sequence C-SEEDHLPLQVLQSH corresponding to C-Terminus according to Uniprot Mouse NP_035836.2. | |
Affinity Purified | |
RUO | |
51393 | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction