Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG1 Antibody (1314D), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB48181Z
Description
VSIG1 Monoclonal antibody specifically detects VSIG1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
VSIG1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
340547 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
1314D | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1 | |
Mouse myeloma cell line, NS0-derived human VSIG-1, Val22-Gly234, Accession # Q86XK7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction