Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG1 Monoclonal Antibody (CT1), Biotin
GREENER_CHOICE

Catalog No. PIMA528799
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528799 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528799 Supplier Invitrogen™ Supplier No. MA528799
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSIG1 Monoclonal Antibody for Flow

VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG1
Applications Flow Cytometry
Classification Monoclonal
Clone CT1
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4
Gene VSIG1
Gene Accession No. Q9PWR4
Gene Alias 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein
Gene Symbols VSIG1
Host Species Mouse
Immunogen Chicken thymocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414795
Target Species Avian, Chicken
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.