Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557606 Supplier Invitrogen™ Supplier No. PA557606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPKPTQEPAP EPAPGSEPMA VPDLDIELEL EPETQSELEP EPEPEPESEP GVVVEPLSED EKGVVK Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.

VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VSIG1
Gene Accession No. Q86XK7
Gene Alias 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein
Gene Symbols VSIG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VSIG1. Recombinant protein control fragment (Product #RP-95433).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.