Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589497 Supplier Invitrogen™ Supplier No. PA589497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASREQLSIQW SFFHKKEMEP ISIYFSQGGQ AVAIGQFKDR ITGSNDPGNA SITISHMQPA DSGIYICDVN NPPDFLGQNQ GILNVSVLVK PSKPLCSVQG RPETGHTISL SCLSALGTPS PVYYWHKLEG RDIVPVKENF NPTTGILVIG NLTNFEQGYY QCTAINRLGN SSCEIDLTSS H; Positive Samples: BxPC3, OVCAR3, Mouse testis, Mouse stomach, Mouse pancreas; Cellular Location: Membrane, Single-pass type I membrane protein.

VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VSIG1
Gene Accession No. Q4KLY3, Q86XK7, Q9D2J4
Gene Alias 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein
Gene Symbols VSIG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-230 of human VSIG1 (NP_8724131).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315920, 340547, 78789
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.