Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584527 Supplier Invitrogen™ Supplier No. PA584527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTCQVSGAYP PAKILWLRNL TQPEVIIQPS SRHLITQDGQ NSTLTIHNCS QDLDEGYYIC RADSPVGVRE MEIWLSVK Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 72%.

VSIG10 (V-Set And Immunoglobulin Domain Containing 10) is a Protein Coding gene. Diseases associated with VSIG10 include Adenoid Hypertrophy. An important paralog of this gene is CNTN5. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VSIG10
Gene Accession No. Q8N0Z9
Gene Alias RGD1565800; V-set and immunoglobulin domain containing 10; V-set and immunoglobulin domain-containing protein 10; Vsig10
Gene Symbols VSIG10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VSIG10. Recombinant protein control fragment (Product #RP-108464).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.