Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSIG10L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566864
Description
Immunogen sequence: PSFTVKTPASN ISTQVSHTKL SVEAPDSKFS PDDMDLKLSA QSPESKFSAE THSAASFPQQ VGGPLAVLVG TTIRLPLVPI PNP Highest antigen sequence identity to the following orthologs - mouse 57%, rat 64%.
VSIG10L is a protein coding gene.
Specifications
| VSIG10L | |
| Polyclonal | |
| Unconjugated | |
| VSIG10L | |
| V-set and immunoglobulin domain containing 10 like; V-set and immunoglobulin domain-containing protein 10-like; VSIG10L; VSIG10-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VR7 | |
| VSIG10L | |
| Recombinant Human VSIG10L. Recombinant protein control fragment (Product #RP-107515). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction