Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG2 Antibody (541522), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB51771C
Description
VSIG2 Monoclonal antibody specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
VSIG2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23584 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
541522 | |
Flow Cytometry, CyTOF-ready | |
2210413P10Rik, Cortical thymocyte-like protein, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession # Q961Q7-1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction