Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG2 Antibody (OTI2D8), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274858MFV450
Description
VSIG2 Monoclonal antibody specifically detects VSIG2 in Human samples. It is validated for Flow CytometrySpecifications
VSIG2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
23584 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI2D8 | |
Flow Cytometry | |
2210413P10Rik, Cortical thymocyte-like protein, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
Full length human recombinant protein of human VSIG2 (NP_055127) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction