Learn More
Invitrogen™ VSIG2 Monoclonal Antibody (OTI1F8)

Description
VSIG2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | VSIG2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1F8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | VSIG2 |
| Gene Accession No. | Q96IQ7 |
| Gene Alias | 1190004B15Rik; 2210413P10Rik; AU040484; cortical thymocyte receptor (X. laevis CTX) like; cortical thymocyte-like protein; CTH; CT-like protein; Ctm; CTX; CTXL; RGD1561274; UNQ2770/PRO7154; V-set and immunoglobulin domain containing 2; V-set and immunoglobulin domain-containing protein 2; VSIG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.