Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VSIG2 Mouse anti-Human, Clone: OTI5A10, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA502161

Description
CTXL, a cortical thymocyte marker, is a member of the immunoglobulin (Ig) superfamily and has features of both antigen specific receptors and adhesion molecules. This single pass, type I protein contains a signal peptide, an extracellular V type Ig like domain followed by a C2 type Ig like domain and a cytoplasmic tail. Expression levels of CTXL are high in the stomach, colon, prostate, trachea, and thyroid gland, with lower levels observed in bladder and lung tissues. CTXL is not expressed in the thymus. CTXL is developmentally regulated and may be involved in thymocyte selection. It localizes to junctional complexes between endothelial and epithelial cells and may play a role in cell-cell adhesion and transmigration of leukocytes.Specifications
VSIG2 | |
Monoclonal | |
0.75 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96IQ7 | |
VSIG2 | |
Full length human recombit protein of human VSIG2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5A10 | |
Unconjugated | |
VSIG2 | |
2210413P10Rik, CTH, CTXL | |
Mouse | |
Affinity Chromatography | |
RUO | |
23584 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction